accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109-94-4

IUPAC Name: ethyl formate
Synonyms: ethyl formate carboxylic acid oxaethane formiato de etilo ethyl formic ester aethylformiat areginal ethyle (formiate d') formic ether ethylformiaat
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: WBJINCZRORDGAQ-UHFFFAOYSA-N
SMILES: CCOC=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (5)
  • (7)
  • (1)
  • (4)