accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109-96-6

IUPAC Name: 2,5-dihydro-1H-pyrrole
Synonyms: 3-pyrroline delta3-pyrroline delta(3)-pyrroline
Molecular Weight (g/mol): 69.11
Molecular Formula: C4H7N
InChi Key: JVQIKJMSUIMUDI-UHFFFAOYSA-N
SMILES: C1NCC=C1
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)