accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1091606-67-5

IUPAC Name: (2-amino-1,2-diphenylethyl)diphenylphosphane
Synonyms: (2-amino-1,2-diphenylethyl)diphenylphosphane
Molecular Weight (g/mol): 381.46
Molecular Formula: C26H24NP
InChi Key: MIPGTOGPLXZBQX-UHFFFAOYNA-N
SMILES: NC(C(P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)