accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFFOHO

CAS RN 1092460-66-6

IUPAC Name: 3-[3-(difluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[3-(difluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 216.18
Molecular Formula: C10H10F2O3
InChi Key: GWTSEGFRVOHJSG-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(OC(F)F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results