accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1092460-69-9

IUPAC Name: 3-(4,5-difluoro-2-methoxyphenyl)propanoic acid
Synonyms: 3-(4,5-difluoro-2-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 216.18
Molecular Formula: C10H10F2O3
InChi Key: WDTSALDVVUTTSK-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(F)C=C1CCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results