accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FNOFFF

CAS RN 1092460-82-6

IUPAC Name: 5-fluoro-2-(trifluoromethoxy)benzonitrile
Synonyms: 5-fluoro-2-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: DIDAOKVLBIJKDX-UHFFFAOYSA-N
SMILES: FC1=CC(C#N)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results