accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1092461-39-6

CH3OFNH2F

CAS RN 1092461-39-6

IUPAC Name: 1-(4-ethoxy-2,6-difluorophenyl)methanamine
Synonyms: 1-(4-ethoxy-2,6-difluorophenyl)methanamine
Molecular Weight (g/mol): 187.19
Molecular Formula: C9H11F2NO
InChi Key: LXXPUAONYYRXSV-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=C(CN)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results