accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 109299-78-7

IUPAC Name: (pyrimidin-5-yl)boronic acid
Synonyms: pyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 123.91
Molecular Formula: C4H5BN2O2
InChi Key: HZFPPBMKGYINDF-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)