accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNOOHHNOO

CAS RN 109425-51-6

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[1-(triphenylmethyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 619.72
Molecular Formula: C40H33N3O4
InChi Key: XXMYDXUIZKNHDT-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results