accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1094411-96-7

CH3H3CCH3NSOHO

CAS RN 1094411-96-7

IUPAC Name: 2-(2-tert-butyl-1,3-thiazol-4-yl)acetic acid
Synonyms: (2-tert-butyl-1,3-thiazol-4-yl)acetic acid
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: AOJJREQRLZDBAA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=NC(CC(O)=O)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results