CAS RN 109461-44-1
IUPAC Name:
2-(4-chlorophenyl)-2-methylmorpholine
Synonyms:
2-(4-chlorophenyl)-2-methylmorpholine
Molecular Weight (g/mol):
211.69
Molecular Formula:
C11H14ClNO
InChi Key:
IJDDASQRAPIORY-UHFFFAOYNA-N
SMILES:
CC1(CNCCO1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
2-(4-Chlorophenyl)-2-methylmorpholine, 99%, Thermo Scientific Chemicals
CAS: 109461-44-1 Molecular Formula: C11H14ClNO Molecular Weight (g/mol): 211.689 MDL Number: MFCD08061115 InChI Key: IJDDASQRAPIORY-UHFFFAOYSA-N Synonym: 2-4-chlorophenyl-2-methylmorpholine,2-4-chloro-phenyl-2-methyl-morpholine,acmc-1c8s6,morpholine,2-4-chlorophenyl-2-methyl PubChem CID: 3066043 IUPAC Name: 2-(4-chlorophenyl)-2-methylmorpholine SMILES: CC1(CNCCO1)C2=CC=C(C=C2)Cl