CAS RN 1095714-91-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Benzyl Epinephrine, TRC
CAS: 1095714-91-2 Molecular Formula: C16 H19 N O3 Molecular Weight (g/mol): 273.33 Synonym: rac Adrenaline EP Impurity D IUPAC Name: 4-[2-[benzyl(methyl)amino]-1-hydroxy-ethyl]benzene-1,2-diol SMILES: CN(CC(O)c1ccc(O)c(O)c1)Cc2ccccc2