accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OOHNNOBr

CAS RN 109770-82-3

IUPAC Name: tert-butyl N-[(3-bromo-4,5-dihydro-1,2-oxazol-5-yl)methyl]carbamate
Synonyms: tert-butyl N-[(3-bromo-4,5-dihydro-1,2-oxazol-5-yl)methyl]carbamate
Molecular Weight (g/mol): 279.13
Molecular Formula: C9H15BrN2O3
InChi Key: AHAHZEKELWWMSJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCC1CC(Br)=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results