accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNFFFOO

CAS RN 109925-42-0

IUPAC Name: 1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Synonyms: 1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 270.21
Molecular Formula: C12H9F3N2O2
InChi Key: JDLPKSIFGPLXAU-UHFFFAOYSA-N
SMILES: CN1N=C(C(C=O)=C1OC1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results