accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 110-00-9

IUPAC Name: furan
Synonyms: furan divinylene oxide tetrole furfuran oxacyclopentadiene rcra waste number U124 axole hsdb 89
Molecular Weight (g/mol): 68.08
Molecular Formula: C4H4O
InChi Key: YLQBMQCUIZJEEH-UHFFFAOYSA-N
SMILES: O1C=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)