accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHO

CAS RN 110-15-6

IUPAC Name: butanedioic acid
Synonyms: succinic acid succinicun acidum kyselina jantarova dicarboxylic acid acidum succinicum acide butanedioique amber acid acide succinique succinic acid (substance) acid, succinic
Molecular Weight (g/mol): 118.09
Molecular Formula: C4H6O4
InChi Key: KDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILES: OC(=O)CCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (25)
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
Percent Purity 
  • (6)
  • (2)
  • (1)
  • (2)
  • (5)
Grade 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (5)
  • (3)
  • (4)
  • (3)
  • (25)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (6)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)