accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3NCH3CH3

CAS RN 110-18-9

IUPAC Name: [2-(dimethylamino)ethyl]dimethylamine
Synonyms: temed tetramethyldiaminoethane tmeda complex tetramethylethylenediamine propamine D tmen tetrameen tetramethyl ethylene diamine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES: CN(C)CCN(C)C
Molecular Weight (g/mol) 
  • (2)
  • (17)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
Physical Form 
  • (23)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (1)
  • (12)
  • (1)
Boiling Point 
  • (4)
  • (5)
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (9)
  • (5)
  • (2)
  • (2)
  • (17)
  • (9)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (23)
  • (2)
  • (3)
  • (2)
  • (1)
  • (12)
  • (1)
  • (4)
  • (5)
  • (9)
  • (5)
  • (6)
  • (3)
  • (3)
  • (1)