accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110-19-0

IUPAC Name: 2-methylpropyl acetate
Synonyms: isobutyl acetate fema number 2175 acetate d'isobutyle isobutylester kyseliny octove isobutylacetat isobutylazetat essigsaeureisobutylester isobutyl acetate (substance) isobutyl acetate (natural)
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: GJRQTCIYDGXPES-UHFFFAOYSA-N
SMILES: CC(C)COC(C)=O
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (6)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)