accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110-46-3

IUPAC Name: 3-methylbutyl nitrite
Synonyms: aspiral amyl nitrite I amyl nitrites ''amyl nitrite'', mixed isomers IPN pentyl nitrite nitrite isopentyl alcohol nitramyl amyl nitrit amyl nitrite(mixed isomers)
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: OWFXIOWLTKNBAP-UHFFFAOYSA-N
SMILES: CC(C)CCON=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (6)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)