accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2

CAS RN 110-58-7

IUPAC Name: pentan-1-amine
Synonyms: amylamine norleucamine
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: DPBLXKKOBLCELK-UHFFFAOYSA-N
SMILES: CCCCCN
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)