accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110-60-1

IUPAC Name: butane-1,4-diamine
Synonyms: putrescine putreszin putrescina
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: KIDHWZJUCRJVML-UHFFFAOYSA-N
SMILES: NCCCCN
Molecular Weight (g/mol) 
  • (7)
  • (6)
Quantity 
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (7)
  • (6)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)