accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNNHCH3

CAS RN 110-70-3

IUPAC Name: methyl[2-(methylamino)ethyl]amine
Synonyms: methyl[2-(methylamino)ethyl]amine
Molecular Weight (g/mol): 88.15
Molecular Formula: C4H12N2
InChi Key: KVKFRMCSXWQSNT-UHFFFAOYSA-N
SMILES: CNCCNC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)