CAS RN 110-77-0
IUPAC Name:
2-(ethylsulfanyl)ethan-1-ol
Synonyms:
ethanol, 2-(ethylthio)-
Molecular Weight (g/mol):
106.18
Molecular Formula:
C4H10OS
InChi Key:
LNRIEBFNWGMXKP-UHFFFAOYSA-N
SMILES:
CCSCCO
1
–
3
of
3
results
2
Ethyl 2-hydroxyethyl sulfide, 98%
CAS: 110-77-0 Molecular Formula: C4H10OS Molecular Weight (g/mol): 106.18 MDL Number: MFCD00002909 InChI Key: LNRIEBFNWGMXKP-UHFFFAOYSA-N Synonym: 2-ethylthio ethanol,ethyl 2-hydroxyethyl sulfide,ethanol, 2-ethylthio,ethyl thioethanol,2-hydroxyethyl ethyl sulfide,ethyl 2-hydroxyethyl thioether,beta-ethylthioethanol,2-ethylsulfanyl ethanol,beta-hydroxydiethyl sulfide,2-ethylthioethanol PubChem CID: 8075 IUPAC Name: 2-ethylsulfanylethanol SMILES: CCSCCO