accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110-83-8

IUPAC Name: cyclohexene
Synonyms: cyclohexene hexanaphthylene tetrahydrobenzene benzene, tetrahydro- benzenetetrahydride tetrahydro-benzene cykloheksen cyclohexene (substance) zyklohexen benzene tetrahydride
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N
SMILES: C1CCC=CC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (6)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (6)
Boiling Point 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (6)
  • (4)
  • (1)