accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)CH3NH2(R)CH3CH3

CAS RN 110-96-3

IUPAC Name: [(2R)-butan-2-yl][(2S)-butan-2-yl]azanium
Synonyms: (2R)-butan-2-yl((2S)-butan-2-yl)azanium
Molecular Weight (g/mol): 130.25
Molecular Formula: C8H20N
InChi Key: OBYVIBDTOCAXSN-OCAPTIKFSA-O
SMILES: CC[C@H](C)[NH2+][C@H](C)CC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)