accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110-99-6

IUPAC Name: 2-(carboxymethoxy)acetic acid
Synonyms: diglycolic acid
Molecular Weight (g/mol): 134.09
Molecular Formula: C4H6O5
InChi Key: QEVGZEDELICMKH-UHFFFAOYSA-N
SMILES: OC(=O)COCC(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results