accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FOHONH2

CAS RN 110117-84-5

IUPAC Name: 2-amino-3-(3-fluorophenyl)propanoic acid
Synonyms: 3-fluorophenylalanine
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: VWHRYODZTDMVSS-UHFFFAOYNA-N
SMILES: NC(CC1=CC(F)=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)