accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1102-19-8

IUPAC Name: 1,1'-dibenzyl-[4,4'-bipyridine]-1,1'-diium dichloride
Synonyms: benzyl viologen dichloride
Molecular Weight (g/mol): 409.35
Molecular Formula: C24H22Cl2N2
InChi Key: NLOIIDFMYPFJKP-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].C(C1=CC=CC=C1)[N+]1=CC=C(C=C1)C1=CC=[N+](CC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results