CAS RN 11029-12-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Enocyanin, TRC
CAS: 11029-12-2 Molecular Formula: C27H31O16 Molecular Weight (g/mol): 611.53