CAS RN 1103522-45-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Despropyl-macitentan, TRC
CAS: 1103522-45-7 Molecular Formula: C16H14Br2N6O4S Molecular Weight (g/mol): 546.19 Synonym: N-[5-(4-Bromophenyl)-6-[2-[(5-bromo-2-pyrimidinyl)oxy]ethoxy]-4-pyrimidinyl]-sulfamide, ACT-132577 SMILES: O=S(N)(NC1=NC=NC(OCCOC2=NC=C(Br)C=N2)=C1C3=CC=C(Br)C=C3)=O