accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOClCl

CAS RN 110763-09-2

IUPAC Name: 5-chloro-1-methyl-1H-pyrazole-4-carbonyl chloride
Synonyms: 5-chloro-1-methylpyrazole-4-carbonyl chloride
Molecular Weight (g/mol): 179
Molecular Formula: C5H4Cl2N2O
InChi Key: XFDCYGXUXLYMNP-UHFFFAOYSA-N
SMILES: CN1N=CC(C(Cl)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results