accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 110802-84-1

ClIrClClClClClHH

CAS RN 110802-84-1

IUPAC Name: tetrachloroiridium hydrate dihydrochloride
Synonyms: tetrachloroiridium hydrate dihydrochloride
Molecular Weight (g/mol): 424.95
Molecular Formula: Cl6H4IrO
InChi Key: YDVQBPXDKJKDME-UHFFFAOYSA-J
SMILES: O.Cl.Cl.Cl[Ir](Cl)(Cl)Cl
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results