accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-01-3

IUPAC Name: 2,6,10,15,19,23-hexamethyltetracosane
Synonyms: squalane caswell no. 482A hexamethyltetracosane dodecahydrosqualene perhydrosqualene cosbiol epa pesticide chemical code 045503 hexamethyl tetracosane robane spinacane
Molecular Weight (g/mol): 422.83
Molecular Formula: C30H62
InChi Key: PRAKJMSDJKAYCZ-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)