accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-02-4

IUPAC Name: (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
Synonyms: squalene squalene (substance) (E,E,E,E)-squalene 角鲨烯 all-trans-squalene spinacen trans-squalene squalen nikko squalane EX spinacene
Molecular Weight (g/mol): 410.73
Molecular Formula: C30H50
InChi Key: YYGNTYWPHWGJRM-AAJYLUCBSA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (6)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)