accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOSOHO

CAS RN 111-17-1

IUPAC Name: 3-[(2-carboxyethyl)sulfanyl]propanoic acid
Synonyms: thiodipropionic acid
Molecular Weight (g/mol): 178.2
Molecular Formula: C6H10O4S
InChi Key: ODJQKYXPKWQWNK-UHFFFAOYSA-N
SMILES: OC(=O)CCSCCC(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)