accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-18-2

H3CNCH3NCH3CH3

CAS RN 111-18-2

IUPAC Name: [6-(dimethylamino)hexyl]dimethylamine
Synonyms: [6-(dimethylamino)hexyl]dimethylamine
Molecular Weight (g/mol): 172.32
Molecular Formula: C10H24N2
InChi Key: TXXWBTOATXBWDR-UHFFFAOYSA-N
SMILES: CN(C)CCCCCCN(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results