accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOClO

CAS RN 111-19-3

IUPAC Name: decanedioyl dichloride
Synonyms: decanedioyl dichloride
Molecular Weight (g/mol): 239.14
Molecular Formula: C10H16Cl2O2
InChi Key: WMPOZLHMGVKUEJ-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCCCCCC(Cl)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)