accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-25-1

IUPAC Name: 1-bromohexane
Synonyms: 1-bromohexane
Molecular Weight (g/mol): 165.07
Molecular Formula: C6H13Br
InChi Key: MNDIARAMWBIKFW-UHFFFAOYSA-N
SMILES: CCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)