accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2

CAS RN 111-26-2

IUPAC Name: hexan-1-amine
Synonyms: hexylamine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: BMVXCPBXGZKUPN-UHFFFAOYSA-N
SMILES: CCCCCCN
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)