accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-39-7

H3CNHNH2

CAS RN 111-39-7

IUPAC Name: (2-aminoethyl)(propyl)amine
Synonyms: (2-aminoethyl)(propyl)amine
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: CFNHVUGPXZUTRR-UHFFFAOYSA-N
SMILES: CCCNCCN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results