accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3

CAS RN 111-47-7

IUPAC Name: 1-(propylsulfanyl)propane
Synonyms: di-N-propylsulfide
Molecular Weight (g/mol): 118.24
Molecular Formula: C6H14S
InChi Key: ZERULLAPCVRMCO-UHFFFAOYSA-N
SMILES: CCCSCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results