accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-65-9

IUPAC Name: octane
Synonyms: octane ottani oktanen n-oktan oktan N-octane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: TVMXDCGIABBOFY-UHFFFAOYSA-N
SMILES: CCCCCCCC
Molecular Weight (g/mol) 
  • (12)
  • (1)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (9)
Grade 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (12)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (9)
  • (2)
  • (4)
  • (3)
  • (3)
  • (6)
  • (9)
  • (1)