accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111-66-0

IUPAC Name: oct-1-ene
Synonyms: 1-octene 1-octene (substance) 1-caprylene caprylene octene (mixture of isomers)
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: KWKAKUADMBZCLK-UHFFFAOYSA-N
SMILES: CCCCCCC=C
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (5)