accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 111-80-8

IUPAC Name: methyl non-2-ynoate
Synonyms: methyl 2-nonynoate methyl octyne carbonate methyl octine carbonate methyloctyne carboxylate
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: NTLJTUMJJWVCTL-UHFFFAOYSA-N
SMILES: CCCCCCC#CC(=O)OC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results