accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3

CAS RN 111-82-0

IUPAC Name: methyl dodecanoate
Synonyms: methyl laurate
Molecular Weight (g/mol): 214.35
Molecular Formula: C13H26O2
InChi Key: UQDUPQYQJKYHQI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (7)