CAS RN 111-84-2
IUPAC Name:
nonane
Synonyms:
nonane N-nonane shellsol 140 nonane (substance)
Molecular Weight (g/mol):
128.26
Molecular Formula:
C9H20
InChi Key:
BKIMMITUMNQMOS-UHFFFAOYSA-N
SMILES:
CCCCCCCCC
1
–
4
of
4
results
2
n-Nonane, 99%, pure
CAS: 111-84-2 MDL Number: MFCD00009574 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
4
n-Nonane 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality