accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNCH3

CAS RN 111-92-2

IUPAC Name: dibutylamine
Synonyms: dibutylamine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: JQVDAXLFBXTEQA-UHFFFAOYSA-N
SMILES: CCCCNCCCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (6)
Physical Form 
  • (2)
  • (8)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (6)
  • (3)
  • (2)
  • (2)
  • (4)
  • (2)