accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 11103-72-3

NH3NH3NH3NH3NH3NH3NH3NH3NH3NH3NH3NH3NH3NH3H2OH2OClClClClClClRuRuRu

CAS RN 11103-72-3

IUPAC Name: tris(λ²-ruthenium(2+)) tetradecaamine dihydrate hexachloride
Synonyms: ruthenium red
Molecular Weight (g/mol): 790.37
Molecular Formula: Cl6H46N14O2Ru3
InChi Key: JQJSTVUROJELSR-UHFFFAOYSA-H
SMILES: N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[Ru++]
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)