CAS RN 1110813-31-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Dacomitinib, TRC
CAS: 1110813-31-4 Molecular Formula: C24 H25 Cl F N5 O2 Molecular Weight (g/mol): 469.94 IUPAC Name: (E)-N-[4-(3-chloro-4-fluoroanilino)-7-methoxyquinazolin-6-yl]-4-piperidin-1-ylbut-2-enamide SMILES: COc1cc2ncnc(Nc3ccc(F)c(Cl)c3)c2cc1NC(=O)\C=C\CN4CCCCC4
2
R&D Systems™ Dacomitinib
A Tocris product, part of R&D Systems. Potent irreversible pan ErbB inhibitor