accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3PCH3ClO

CAS RN 1111-92-8

IUPAC Name: dimethylphosphinoyl chloride
Synonyms: dimethylphosphinoyl chloride
Molecular Weight (g/mol): 112.49
Molecular Formula: C2H6ClOP
InChi Key: CVNMBKFJYRAHPO-UHFFFAOYSA-N
SMILES: CP(C)(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)